Learn More
BrandTech™ VITLAB™ Polypropylene Pipet Fillers
Click to view available options
For Use With (Equipment):
Pipettes, 10 mL
Pipettes, 25 mL
Color:
Green
Red
Unit Size:
Each
Description
- For pipetting liquids
- Slow rotation of the actuator-wheels draws liquid into the pipette
- Pressing the air bleed valve automatically empties the pipette without returning the piston
Specifications
Specifications
| Material | Polypropylene |
| For Use With (Application) | Pipetting liquids |
| Quantity | 1 Each |
| Color | Green |
| Type | Pipette Filler |
| For Use With (Equipment) | Pipettes, 10 mL |