Learn More
BRAND™ Polypropylene Photographic Trays
Product Code. p-8010318 Shop All Brand Products
Click to view available options
Height (Metric):
45 mm
75 mm
120 mm
140 mm
160 mm
Unit Size:
Each
Product Code. 10643782
Brand: BRAND™ 156640
Only null left
Add to basket
Add to basket
White stackable polypropylene (PP) photographic trays with ribbed bottom that improves stability.
  • White stackable polypropylene (PP) photographic trays
  • Ribbed bottom improves stability, reinforced rim, slightly sloping walls
  • Autoclavable (121°C)
TRUSTED_SUSTAINABILITY
Specifications
Color White
Height (Metric) 45 mm
Type Photographic Tray
Includes Ribbed bottom, reinforced rim
Shape Rectangular, stackable
Autoclavable Autoclavable (121°C)