Learn More
Kimble Chase™ Polypropylene Open-Top Closure
Product Code. 11893033
Click to view available options
Color:
Blue
White
Unit Size:
Pack of 1000
Product Code. 11893033
Brand: Kimble Chase™ 73813WH13425
Only null left
Specifications
Color White
Material Silicone, Polypropylene, PTFE
Pre-slit Yes
Septum Red Silicone
Quantity 1000 Pk.
Type Screw Cap
Closure Size 13-425