Learn More
Brady™ Polypropylene Label Tape for BradyJet J4000
Product Code. 17469222
Click to view available options
Width (Metric):
57.1 mm
101.6 mm
152.4 mm
203.2 mm
Unit Size:
Each
Product Code. 17469222
Brand: Brady™ 152785
Only null left
Add to basket
Add to basket
This B-7425J polypropylene material is ideal for GHS and other full-color identification applications where resistance to harsh chemicals is required.
  • Matte white polypropylene with acrylic adhesive is Inkjet printable
  • Low-cost, full-color GHS labeling option withstands chemical exposure
  • Service temperature of -80°C to 70°C
  • An ink cartridge is required to print this material
Specifications
Autoclavable Autoclavable
Certifications/Compliance Compliant with Directive 2011/65/EU and 2015/863
Adhesive Type Acrylic, Permanent
Coating None
Color White
Finish Matte
Format Continuous
Material Polypropylene (Inkjet-receptive)
Printable Sides Single-side Printable
Print Technology Ink Jet
Product Type Label
Sterility Non-sterile
For Use With (Application) Hazard Communication Identification, Equipment Labeling, Laboratory Labeling, General Identification
For Use With (Equipment) BradyJet J4000 Color Label Printer
Labels per Row 1
Min. Operating Temperature (Metric) -80°C
No. per Pack 1 Tape/Bag
Quantity 1 Tape
Temperature Range (English) -112°F to +158°F
Temperature Range (Metric) -80°C to +70°C
Thickness (English) 0.005 in.
Thickness (Metric) 0.13 mm
Dimensions (LxW) 30.48 m x 152.4 mm
Length (English) 100 ft.
Length (Metric) 30.5 m
Width (English) 6 in.
Width (Metric) 152.4 mm
Alternate Part Number J40C-6000-7425J