Learn More
Brady™ Polypropylene: ISO Safety Sign - Eyewash station
Sign compliant to ISO 7010.
Brand:  Brady™ 815558
Additional Details : Weight : 0.03200kg
 View more versions of this product
Product Code. 15624337
  • 9.45 EUR / Each
Estimated Shipment: 30-05-2024
Description
  • Polypropylene
  • For indoor and outdoor use
Specifications
Specifications
ISO 7010
1 Ea.
148 mm
148 mm
Square
White on Green
Polypropylene
Indoor and Outdoor
1.501 mm
-40°C to 120°C
Global