Learn More
Kappes Systeme™ Polypropylene Euro Transport Box
Product Code. 10004132
Change view
Click to view available options
Color:
Blue
Gray
Red
Dimensions (L x W x H):
400 x 300 x 120 mm
400 x 300 x 180 mm
400 x 300 x 220 mm
600 x 400 x 120 mm
600 x 400 x 180 mm
600 x 400 x 220 mm
600 x 400 x 320 mm
600 x 400 x 420 mm
Unit Size:
Each
22 product options available for selection
| Product Code. | Color | Dimensions (L x W x H) | unitSize |
|---|---|---|---|
| 10004132 | Red | 600 x 400 x 320 mm | Each |
| 10093952 | Blue | 400 x 300 x 120 mm | Each |
| 10522943 | Blue | 400 x 300 x 220 mm | Each |
| 10368872 | Blue | 400 x 300 x 180 mm | Each |
| 10125722 | Blue | 600 x 400 x 120 mm | Each |
| 10208302 | Blue | 600 x 400 x 180 mm | Each |
| 10309312 | Blue | 600 x 400 x 220 mm | Each |
| 10238012 | Blue | 600 x 400 x 320 mm | Each |
| 10512563 | Blue | 600 x 400 x 420 mm | Each |
| 10247722 | Gray | 400 x 300 x 120 mm | Each |
| 10389062 | Gray | 400 x 300 x 220 mm | Each |
| 10635643 | Gray | 600 x 400 x 180 mm | Each |
| 10710814 | Gray | 600 x 400 x 220 mm | Each |
| 10155482 | Gray | 600 x 400 x 320 mm | Each |
| 10248012 | Gray | 600 x 400 x 420 mm | Each |
| 10532943 | Red | 400 x 300 x 220 mm | Each |
| 10768503 | Red | 400 x 300 x 120 mm | Each |
| 10207822 | Red | 400 x 300 x 180 mm | Each |
| 10730234 | Red | 600 x 400 x 120 mm | Each |
| 10700814 | Red | 600 x 400 x 180 mm | Each |
| 10615823 | Red | 600 x 400 x 220 mm | Each |
| 10412722 | Red | 600 x 400 x 420 mm | Each |
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 22 options available.
This item is not returnable.
View return policy
This item is not returnable.
View return policy
Specifications
Specifications
| Type | Transport Bin |
| Material | Polypropylene |
| Color | Red |
| Shape | Rectangular |
| For Use With | Conveyor, Grabber Technology, Storage and Retrieval Machine |
| For Use With (Application) | Food Safe |
| Quantity | 1/Pk. |
| Dimensions (L x W x H) | 600 x 400 x 320 mm |
| Length (Metric) | 600 mm |
| Width (Metric) | 400 mm |
| Height (Metric) | 320 mm |
| Show More Show Less |