Learn More
JG Finneran Associates™ Polypropylene Convenience Pack With Shell Vials
Click to view available options
Volume (Metric):
1 mL
2 mL
Color:
Amber
Clear
Unit Size:
Pack of 100
Specifications
Specifications
| Color | Amber |
| Material | Polypropylene |
| Volume (Metric) | 1 mL |
| Includes | Clear Shell Vial and Plugs |