Learn More
Kartell™ Polypropylene Conical Erlenmeyer Flasks
Translusent in design to allow one to view the mixture with ease. Kartell™ Polypropylene Conical Erlenmeyer Flasks are a simple flask that has a variety of applications in both chemistry and biology.
Brand: Kartell™ 0146400
Description
Translusent in design to allow one to view the mixture with ease. Kartell™ Polypropylene Conical Erlenmeyer Flasks are a simple flask that has a variety of applications in both chemistry and biology.
- For general laboratory use
- Provided with standardized neck
- Autoclavable
- Suitable for foodstuff
Specifications
| Translucent | |
| Polypropylene | |
| 29/32 NS | |
| Autoclavable |
| 1000 mL | |
| 50 mL | |
| Conical | |
| Yes |