Learn More
DWK Life Sciences Kimble Chase™ Polypropylene Closure with Ultrasonically Welded PTFE Liner
Description
- Provides superior closure system capable of maintaining its liner through multiple autoclavable operations
- PTFE-coated surface of exposed liner is chemical-resistant and compatible with sensitive, corrosive and volatile solvents
Specifications
Specifications
| Closure Size | 15-415 |
| Liner Material | PTFE, Rubber |
| Product Type | Screw Cap |
| Autoclavable | Autoclavable |
| Material | Polypropylene |
| For Use With (Application) | For repeated autoclaving, to provide superior closure system |
| For Use With (Equipment) | EPA Water Analysis Vials, Screw Thread Culture and Centrifuge Tubes |