Learn More
Macherey-Nagel™ Polypropylene Chromabond Column
71.00 EUR - 188.00 EUR
Product Code | Brand | Column Capacity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Column Capacity | Price | Quantity & Availability | |||||
10082941
|
Macherey-Nagel™
730487 |
6 mL |
71.00 EUR
Pack of 30 |
Estimated Shipment: 04-06-2024
Log in to see stock. |
|||||
10401862
|
Macherey-Nagel™
730507 |
70 mL |
188.00 EUR
Pack of 30 |
Estimated Shipment: 08-05-2024
Log in to see stock. |
|||||
Specifications
Column | |
30 Pack | |
Polypropylene |