Learn More
BRAND™ Polypropylene Bucket
Reinforced rim and handle offer sturdy control. Spout provides easy pouring and prevents spilling.
33.95 EUR - 36.70 EUR
Specifications
Material | Polyproyplene |
---|---|
Color | Translucent |
Handle Material | Polypropylene |
Includes | Spout, reinforced handle |
Quantity | 1 Ea. |
Product Code | Brand | Capacity (Metric) | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Capacity (Metric) | Price | Quantity & Availability | |||||
10395561
|
BRAND™
72376 |
12 L |
33.95 EUR
Each |
Estimated Shipment: 30-04-2024
Log in to see stock. |
|||||
10111871
|
BRAND™
72378 |
15 L |
36.70 EUR
Each |
Estimated Shipment: 26-04-2024
Log in to see stock. |
|||||
Description
- Polypropylene construction
- Reinforced rim and handle
- Includes spout
- Lid not included
Specifications
Polyproyplene | |
Polypropylene | |
1 Ea. |
Translucent | |
Spout, reinforced handle |