Learn More
Masterflex™ Polypropylene Barbed Fitting
Product Code. 11979658
Click to view available options
For Use With:
1/2 NPT x 1/2 in. tubing I.D.
1/2 NPT x 1/4 in. tubing I.D.
1/4 NPT x 1/2 in. tubing I.D.
1/4 NPT x 3/8 in. tubing I.D.
3/8 NPT x 1/2 in. tubing I.D.
3/8 NPT x 1/4 in. tubing I.D.
3/8 NPT x 3/8 in. tubing I.D.
Unit Size:
Pack of 10
Product Code. 11979658
Brand: Masterflex™ 4061046
Only null left
Add to basket
Add to basket
Adapts from a NPT male thread to a hose barb. Cole-Parmer™ Polypropylene Barbed Fittings are available in a variety of configurations to meet your needs.
  • Quick and easy assembly
  • Available in packs of ten
Specifications
Material Polypropylene
For Use With 1/2 NPT x 1/2 in. tubing I.D.
Packaging Quantity 10 Pack
Port Size 11/32 in. orifice; 2-3/32 x 1 in.
Thread Size 1/2 in. NPT
Includes ETO or gamma sterilization
Product Type Barbed Fitting