Learn More
Poulten & Graf GmbH Polypropylene Adaptor for FORTUNA™ Dispenser with Ground Joint
Product Code. p-7116396
Click to view available options
Thread Size:
TS 19
TS 24
TS 29
TS 45
TS 60
Unit Size:
Each
Product Code. 10094790
Brand: Poulten & Graf GmbH 101 09245
Only null left
Add to basket
Add to basket
Complete a FORTUNA™ Dispenser with an adapter that comes in a wide selection of threads. Poulten & Graf GmbH Polypropylene Adaptor for FORTUNA™ Dispenser with Ground Joint enables smaller containers to be supported on top the existing reservior.
  • Suitable for OPTIFIX and POLYFIX dispenser
  • Made of Polypropylene
  • Adapter with ground joint
  • Can be used for most chemicals and liquids
Specifications
Product Type Adapters for OPTIFIX and POLYFIX Dispensers
Autoclavable Autoclavable
Color White
Description Adapters for OPTIFIX and POLYFIX dispensers
Material Polypropylene
For Use With (Equipment) OPTIFIX and POLYFIX dispensers
Quantity 1
Thread Size TS 45
Thread Type Ground joint