Learn More
Poulten & Graf GmbH Polypropylene Adaptor for FORTUNA™ Dispenser with Ground Joint
Click to view available options
Thread Size:
TS 19
TS 24
TS 29
TS 45
TS 60
Unit Size:
Each
Description
- Suitable for OPTIFIX and POLYFIX dispenser
- Made of Polypropylene
- Adapter with ground joint
- Can be used for most chemicals and liquids
Specifications
Specifications
| Product Type | Adapters for OPTIFIX and POLYFIX Dispensers |
| Autoclavable | Autoclavable |
| Color | White |
| Description | Adapters for OPTIFIX and POLYFIX dispensers |
| Material | Polypropylene |
| For Use With (Equipment) | OPTIFIX and POLYFIX dispensers |
| Quantity | 1 |
| Thread Size | TS 19 |
| Thread Type | Ground joint |