missing translation for 'onlineSavingsMsg'
Learn More

Polyphosphoric acid, ≥83% phosphate (as P2O5) basis, Honeywell Fluka™

Product Code. 15680700 Shop All Honeywell Fluka Products
Click to view available options
Quantity:
500 g
1 kg
2.5 kg
Unit Size:
1kg
2.5kg
500g
This item is not returnable. View return policy

Product Code. 15680700

Brand: Honeywell-Fluka 041012.5KG

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Chemical Identifiers

CAS 8017-16-1
Molecular Formula (HO3P)n
MDL Number MFCD00084480
InChI Key NBIIXXVUZAFLBC-UHFFFAOYSA-N
IUPAC Name Polyphosphoric acid
SMILES [H]OP(O)(O)=O

Specifications

CAS Max % ≤100%
Quantity 2.5 kg
Percent Purity >83%
Assay of phosphate as P2O5
Chemical Name or Material Polyphosphoric Acid, phosphate (as P2O5) ≥83%
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.