Learn More
Polyoxyl 35 Castor Oil, TRC
Product Code. 30426433
Change view
Click to view available options
Quantity:
100 mg
1 g
50 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30426433 1 g
30420144 100 mg
30422198 50 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 30426433 Supplier Toronto Research Chemicals Supplier No. TRCP6893951G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 61791-12-6
Synonym 1161X, AQ 250, Acconon CA 15, Actinol CS 40, Agnique CSO 30, Alkamuls OR 36, Arlatone 285, Atlas G 1281, Berol 199, Bio-Soft HR 40, Blaunon BR 410, C 125, CO 10, Castor oil polyoxyethylene ether, Cemulsol BR, Chemax CO, Cremophor ELP, EL 30 (emulsifier), Emalex C 50, Emulan ELH, Eumulgin PRT 33, Eumulgin RH 40, Eumulgin RO 40, Eumulgin RT 40, G 1300, GS 2, GS 2 (surfactant), HCO 505, Hairikku AQ 250, Hedipin R 200, Heliwet EO 33, Hextoxide C 15, Hyric AQ 120, Hyric AQ 250, Incrocas 30, Incrocas 35, Incrocas 40, KECAS 40, Kolliphor EL, Kolliphor ELP, Koremul CO 17, Leunapon ER 40, Lipal 200C, Lumulse CO 25, Lumulse CO 40, Lyscil 25C, Mapeg CO 8, Mergital EL 33, Mulgoven EL 719, Mulsifan RT 163, Nalco 2395, Newcol 1500S, Newkalgen D 212, Newpol CZ 300, Nikkol CO, Peregal C 125, Pionin D 212, Press-Wet OC 20A, Protachem CA 30, RT 40, Rhodasurf 870, Ricinox 80, Rokacet R 40, Simulsol 1285, Slovasol EL, Soprophor B, Sorpol CA 20, Sorpol CA 42, Sunaptol CA 350, Surfactol 318, Surfom R 400, Surfonic CO 36, Synlube 106, TRYLOX CO 40, Tagat R, Tanemul KS, Teric 380, Termul 1284, Texofor D 30, Tipal 52C, Toxanon AH, Toximul 8240, Trylox 5900, UR 34, Ukanil 2507, Unitop FFT 40, Vibsol EL 40, Visco L 357, Wettol EM 31, Witconol 5909
SMILES O=C(OCCOCC(COCCOC(CCCCCCC/C=C\CC(O)CCCCCC)=O)OCCOC(CCCCCCC/C=C\CC(CCCCCC)O)=O)CCCCCCC/C=C\CC(O)CCCCCC
Specifications
Quantity 1 g
Chemical Name or Material Polyoxyl 35 Castor Oil