Learn More
Kautex™ Polyethylene Wide Mouth Square Bottles
Click to view available options
Capacity (Metric):
250 mL
500 mL
1000 mL
Diameter (Metric) Inner Neck:
30.8 mm
39.2 mm
Unit Size:
Each
Pack of 164
Pack of 294
Pack of 90
Description
- Easy filling, emptying and cleaning
- Ribbed sides for a secure grip
- Rectangular labelling field
- Rectangular shape optimizes space utilisation
- Leakproof
Specifications
Specifications
| Capacity (Metric) | 1000 mL |
| Color | Natural |
| Diameter (Metric) Inner Neck | 39.2 mm |
| For Use With (Application) | Packaging, Storage, Transport |
| Material | HDPE |
| No. per Case | 90 |
| Quantity | 90/Cs. |
| Shape | Square |
| Height (Metric) | 178.5 mm |
| Product Type | Wide Neck Bottle |