Description
- Enables quick, at-a-glance identification of bottle contents
- Choose from Natural, Red, Yellow, Blue or Green closures
- Bulk quantities available by special order
Specifications
Specifications
| Capacity (Metric) | 250 mL |
| Capacity (English) | 8 oz. |
| Color | Natural |
| Diameter (Metric) Outer Neck | 38 mm |
| Material | Polyethylene, Polypropylene |
| Sterility | Non-sterile |
| Closure Color | Natural |
| Closure Included | Yes |
| Disposable | Reusable |
| Graduated | No |
| Show More |