Learn More
GUERIN PLASTIQUES Polyethylene Sleeves
Product Code. 11896513
Change view
Click to view available options
Size:
0.1 Thick x 100Wmm
0.1 Thick x 120Wmm
0.1 Thick x 140Wmm
0.1 Thick x 160Wmm
0.1 Thick x 180Wmm
0.1 Thick x 200Wmm
0.1 Thick x 250Wmm
0.1 Thick x 300Wmm
0.1 Thick x 350Wmm
0.1 Thick x 55Wmm
0.1 Thick x 64Wmm
Unit Size:
Each
11 product options available for selection
Product selection table with 11 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Size unitSize
11896513 0.1 Thick x 64Wmm Each
11836523 0.1 Thick x 160Wmm Each
11806523 0.1 Thick x 100Wmm Each
11856523 0.1 Thick x 200Wmm Each
11846543 0.1 Thick x 300Wmm Each
11866523 0.1 Thick x 250Wmm Each
11856543 0.1 Thick x 350Wmm Each
11816523 0.1 Thick x 120Wmm Each
11826523 0.1 Thick x 140Wmm Each
11846523 0.1 Thick x 180Wmm Each
11816503 0.1 Thick x 55Wmm Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 11 options available.
11 options
Product Code. 11896513 Supplier GUERIN PLASTIQUES Supplier No. 50150
Only null left
Add to basket
Add to basket
Use for food handling and general applications. GUERIN PLASTIQUES Polyethylene Sleeves are made of transparent, 0.1mm thick polyethylene.
  • 350m long in total
  • Available in a variety of widths
Specifications
Quantity 1/Pk.
Type Sleeve
Material Polyethylene
For Use With (Equipment) Food Quality
Length (Metric) 350 m
Size 0.1 Thick x 64Wmm
Color Transparent