Learn More
Kimberly-Clark™ Kleenex™ Waterproof Pads
Click to view available options
Dimensions (L x W):
500 x 370 mm
Unit Size:
Pack of 4
Description
- Manufactured from polyethylene
- Size: 37.5 x 50 cm
Specifications
Specifications
| Product Type | Tissue |
| Length (English) | 19.7 in. |
| Length (Metric) | 500 mm |
| Width (English) | 14.8 in. |
| Width (Metric) | 375 mm |
| Material | Polyethylene |
| Color | Green |
| Dimensions (L x W) | 500 x 370 mm |
| For Use With (Application) | Used as an underlay in the workplace where liquids are used. |
| Packaging Quantity | 400 Pk. |