Learn More
Asecos™ Polyethylene Sump Pallet
Shop All asecos Products
Click to view available options
Dimensions:
1235 x 1235 x 300 mm
Unit Size:
Each
Description
- Accessory item
- With PE grid
- Storage capacity for 4x 200 L drums
Specifications
Specifications
| For Use With (Equipment) | 4x 200 L drums |
| Dimensions | 1235 x 1235 x 300 mm |
| Weight (Metric) | 33 Kg |