Learn More
Polyethylene, chlorinated
Product Code. 11319648
Click to view available options
Quantity:
250 g
1 kg
Unit Size:
1kg
250g
Product Code. 11319648
Brand: Thermo Scientific Alfa Aesar 044146.30
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
CAS: 64754-90-1
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 64754-90-1
MDL Number MFCD00084424
Specifications
Color White
Physical Form Powder
Solubility Information Soluble in cyclohexanone and toluene
Quantity 250 g
Refractive Index 1.22
Chemical Name or Material Polyethylene, Chlorinated