Learn More
BRAND™ HDPE Bucket
Product Code. p-8010279 Shop All Brand Products
Click to view available options
Capacity (Metric):
5 L
Height (Metric):
240 mm
Unit Size:
Each
Product Code. 10613972
Brand: BRAND™ 71772
Only null left
Add to basket
Add to basket
Manufactured from High-density Polyethylene.BRAND™ HDPE Buckets include a handle, and graduation marks. Without spout. Push-on lid available for separate purchase.
  • Manufactured from High-density Polyethylene.
  • Graduated.
  • Includes handle.
  • Without spout.
  • Push-on lid available for separate purchase.
TRUSTED_SUSTAINABILITY
Specifications
Capacity (Metric) 5 L
Graduations Yes
Height (Metric) 240 mm
Includes Handle