Learn More
Brady™ Polyester Labels for Thermal Transfer Printers
GREENER_CHOICE
Product Code. 12646045
Click to view available options
Size (Metric):
1.27 x 4.57 cm (W x H)
1.52 x 6.67 cm (W x H)
1.9 x 4.57 cm (W x H)
1.9 x 5.59 cm (W x H)
2.54 x 1.9 cm (W x H)
2.54 x 2.54 cm
6.35 W x 6.35 cm H
Finish:
Matte
For Use With (Equipment):
BBP11, BBP12, THT 25 mm Core Printers
BBP81, i5100, i6100, i7100, i7500, PR Plus, IP Series, THT 76 mm Core Printers
Quantity:
1500/Pk.
2500/Pk.
5000/Pk.
Unit Size:
1 roll
Pack of 2500
Product Code. 12646045
Brand: Brady™ 116386
Only null left
Add to basket
Add to basket
For barcoding and identification purposes
  • Available with gloss or matte finish
  • Available in different widths and roll lengths
Specifications
Autoclavable Autoclavable
Certifications/Compliance RoHS 2002/95/EC
Description Thermal Transfer Printable Labels
Adhesive Type Acrylic, Permanent
Coating None
Color White, Translucent
Finish Matte
Format Label
Material Polyester
Printable Sides Single-side Printable
Print Technology Thermal Transfer
Product Type Label
Sterility Non-sterile
For Use With (Application) Laboratory identification such as vials, centrifuge tubes, test tubes, straws, and slides
For Use With (Equipment) BBP11, BBP12, THT 25 mm Core Printers
Labels per Row 1
Min. Operating Temperature (Metric) -196°C
Model THT-122-461-1.5-SC
No. per Pack 1500 Label(s) / Roll
Quantity 1500/Pk.
Shape Rectangle
Size 45.72 mm (W) x 12.70 mm (H)
Size (Metric) 1.27 x 4.57 cm (W x H)
Temperature Range (English) -321°F to +230°F
Temperature Range (Metric) -196°C to +110°C
Thickness (English) 0.0019 in.
Thickness (Metric) 0.048 mm
Vertical Repeat (Metric) 15.88 mm
Dimensions (LxW) 45.72 mm (W) x 12.70 mm (H)
Length (English) 1.8 in.
Width (English) 0.5 in.
Width (Metric) 12.70 mm
Width (English) Printable Area 45.72 mm
Width (Metric) Printable Area 19.05 mm
Height (English) 1.8 in.
Height (English) Printable Area 0.5 in.
Height (Metric) 4.57 cm
Height (Metric) Printable Area 19.05 mm
Diameter (English) 1 in.
Diameter (Metric) 2.54 cm
Alternate Part Number THT-122-461-1.5-SC