Learn More
Serva Electrophoresis™ Polyacryl-amide Servalyt Precotes
Product Code. p-8010999
Encompass_Preferred
Click to view available options
Length (Metric):
125 mm
Separation Range:
pH 3 to 10
pH 3 to 6
Unit Size:
Each
Product Code. 11457883
Brand: Serva Electrophoresis™ 4287402
Specifications
Gel Percentage 5%
Gel Thickness 0.65 mm
Gel Type IEF
Product Type Homogeneous Page
Separation of Protein
Separation Range pH 3 to 6
Quantity 5/Pk.
Length (Metric) 125 mm
Length (Metric) Gel 125 mm
Width (Metric) 125 mm
Width (Metric) Gel 125 mm
Show More Show Less
compliance-icons
Product Identifier
  • SERVALY PRECOTES Range pH 3 - 6
Signal Word
  • Danger
Hazard Category
  • Germ cell mutagenicity Category 1B
  • Carcinogenicity Category 1B
Hazard Statement
  • H340-May cause genetic defects.
  • H350-May cause cancer.
Precautionary Statement
  • P201-Obtain special instructions before use.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P202-Do not handle until all safety precautions have been read and understood.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
  • P405-Store locked up.
  • P501-Dispose of contents/container to .
Supplemental information
  • MIXTURE LIST-Contains : acrylamide