missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Poly-L-Lysine Hydrobromide [MW 4,000-15,000], MP Biomedicals
Shop All MP Biomedicals Products
Click to view available options
Quantity:
25 mg
100 mg
500 mg
Unit Size:
100mg
25mg
500mg
Specifications
Specifications
| Quantity | 25 mg |
| Formula Weight | 4,000 to 15,000 |
| Chemical Name or Material | Poly-L-Lysine Hydrobromide |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction