missing translation for 'onlineSavingsMsg'
Learn More

Poly-DL-alanine, 95%, MP Biomedicals™

Product Code. 11408330 Shop All MP Biomedicals Products
Click to view available options
Quantity:
250 mg
Unit Size:
250mg
This item is not returnable. View return policy

Product Code. 11408330

Brand: MP Biomedicals 0210267783

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Chemical Identifiers

CAS 25281-63-4
Molecular Formula C3H7NO2
MDL Number MFCD00166369

Specifications

Color White
Assay Percent Range 95%
Quantity 250 mg
Formula Weight 1000 to 5000
Physical Form Powder
Chemical Name or Material Poly-DL-alanine
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.