missing translation for 'onlineSavingsMsg'
Learn More

Poly(acrylic acid), 50 wt.% solution in water; approx. M.W. 3,000 to 5,000

Product Code. p-185682 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
5 g
250 g
1 kg
Packaging:
Glass bottle
Unit Size:
1kg
250g
5g
This item is not returnable. View return policy

Product Code. 10696382

Brand: Thermo Scientific Acros 185000050

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 9003-01-4 | (C3H4O2)n | 72.06 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 9003-01-4, 7732-18-5
Molecular Formula (C3H4O2)n
Molecular Weight (g/mol) 72.06
MDL Number MFCD00084394
InChI Key NNMHYFLPFNGQFZ-UHFFFAOYSA-M
Synonym acrylic acid, 2-propenoic acid, propenoic acid, vinylformic acid, acroleic acid, propene acid, ethylenecarboxylic acid, polyacrylate, propenoate, carbomer
PubChem CID 6581
ChEBI CHEBI:18308
SMILES OC(=O)C(-*)C-*

Specifications

Physical Form Liquid
Boiling Point 100.0°C
Linear Formula [-CH2CH(CO2H)-]n
Density 1.1800g/mL
Solubility Information Solubility in water: soluble. Other solubilities: soluble in methanol, ethanol, dioxane
Quantity 5 g
IUPAC Name Poly(acrylic acid)
Percent Purity 48 to 52% (solids)
Infrared Spectrum Authentic
Packaging Glass bottle
Specific Gravity 1.18
Chemical Name or Material Poly(acrylic acid), 50 wt. % Solution in Water
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.