missing translation for 'onlineSavingsMsg'
Learn More

Poloxime, MedChemExpress

Product Code. 16464117
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
500 mg
1 g
Unit Size:
100mg
10mg
1g
500mg
50mg
5mg
This item is not returnable. View return policy

Product Code. 16464117

Brand: Medchem Express HY77195

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Poloxime, a hydrolysis product of poloxin, is a non-ATP-competitive Plk1 inhibitor, with moderate Plk1 inhibitory activity.

Poloxime, a hydrolysis product of poloxin, is a non-ATP-competitive Plk1 inhibitor, with moderate Plk1 inhibitory activity.

Specifications

Chemical Name or Material Poloxime
Physical Form Solid
CAS 17302-61-3
Color Yellow
Quantity 50 mg
Purity Grade Notes Research
Molecular Formula C10H13NO2
For Use With (Application) Cancer-Kinase/protease
Solubility Information DMSO : ≥ 100 mg/mL (557.97 mM) ∣H2O : < 0.1 mg/mL (insoluble)
SMILES CC1=CC(C(C(C)C)=C/C1=N\O)=O
Molecular Weight (g/mol) 179.22
Formula Weight 179.22
Percent Purity 95.0%
Grade Research
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.