missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ POLL Polyclonal Antibody
GREENER_CHOICE

Product Code. 16569871
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16569871 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16569871 Supplier Invitrogen™ Supplier No. PA597040

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Immunogen sequence: GKATCGDVDV LITHPDGRSH RGIFSRLLDS LRQEGFLTDD LVSQEENGQQ QKYLGVCRLP GPGRRHRRLD IIVVPYSEFA CALLYFTGSA HFNRSMRALA KTKGMSLSEH ALSTAVVRNT HGCKVGPGRV LPTPTEKDVF RLLGLPYREP AERDW; Positive Samples: MCF7, HL-60, A-549, NCI-H460, Rat liver; Cellular Location: Nucleus.

The protein encoded by this gene functions as a cofactor for the stimulation of transcriptional elongation by HIV-1 Tat, which binds to the HIV-1 promoter through Tat-TAR interaction. This protein may also serve as a dual-function factor to couple transcription and splicing and to facilitate their reciprocal activation. Alternatively spliced transcript variants have been found for this gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen POLL
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 1.57 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene Poll
Gene Accession No. Q5RKI3, Q9QXE2, Q9UGP5
Gene Alias 1110003P06Rik; AV007317; BETAN; DNA polymerase beta-2; DNA polymerase beta-N; DNA polymerase kappa; DNA polymerase lambda; DNA-directed DNA polymerase lambda; EC 2.7.7.7; FLJ46002; OTTHUMP00000020323; Pol beta2; Pol Lambda; Polk; POLKAPPA; POL-KAPPA; Poll; polymerase (DNA directed), lambda; polymerase (DNA) lambda; RP11-529I10.1
Gene Symbols Poll
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 421-575 of human POLL (NP_0374061).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 27343, 361767, 56626
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.