missing translation for 'onlineSavingsMsg'
Learn More

POGK Rabbit anti-Human, Polyclonal, Proteintech

Product Code. 16843395 Shop All Proteintech Products
Change view
Click to view available options
Quantity:
20 μL
150 μL
Unit Size:
150µL
20µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16843395 20 μL 20µL
16833395 150 μL 150µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16843395 Supplier Proteintech Supplier No. 248881AP20UL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

This gene encodes a 134 kDa protein named strumpellin that is predicted to have multiple transmembrane domains and a spectrin-repeat-containing domain. This ubiquitously expressed gene has its highest expression in skeletal muscle. The protein is named for Strumpell disease; a form of hereditary spastic paraplegia (HSP). Spastic paraplegias are a diverse group of disorders in which the autosomal domit forms are characterized by progressive, lower extremity spasticity caused by axonal degeneration in the terminal portions of the longest descending and ascending corticospinal tracts. More than 30 loci (SPG1-33) have been implicated in hereditary spastic paraplegia diseases.

Specifications

Antigen POGK
Applications Immunofluorescence, Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.26 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
Gene POGK
Gene Accession No. Q9P215
Gene Alias BASS2, KIAA1513, KIAA15131, LST003, POGK, SLTP003
Gene Symbols POGK
Host Species Rabbit
Immunogen POGK Fusion Protein Ag21681
Purification Method Antigen Affinity Chromatography
Quantity 20 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 57645
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.