missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ PODN Polyclonal Antibody

Goat Polyclonal Antibody
Brand: Invitrogen™ PA547446
This item is not returnable.
View return policy
Description
In direct ELISAs and Western blots, approximately 5% cross-reactivity with recombinant human Podocan is observed. Reconstitute at 0.2 mg/mL in sterile PBS.
Non-collagenous proteins of the small leucine-rich repeat (SLR) protein family are considered important components of the extracellular matrix. The extracellular matrix plays an integral role in the pivotal processes of development, tissue repair and metastasis by regulating cell proliferation, differentiation, adhesion and migration. Podocan, also known as PCAN, SLRR5A or PODN, is a 613 amino acid secreted protein belonging to the small leucine-rich proteoglycan (SLRP) family and the class V subfamily. Expressed in kidney, heart, liver, pancreas and vascular smooth muscle cells, Podocan exists as three alternatively spliced isoforms containing twenty LRR (leucine-rich repeats), a unique N-terminal cysteine-rich cluster pattern and a highly acidic C-terminal domain. Podocan is considered a glycoprotein containing N-linked oligosaccharides that may be involved in growth regulation in cardiovascular tissues. Encoded by a gene located on human chromosome 1, Podocan interacts type I collagen (COL1).
Specifications
| PODN | |
| Polyclonal | |
| Unconjugated | |
| Podn | |
| 9430070G18; Pcan; Podn; Podocan; podocan proteoglycan; PRO332; SLRR5A; UNQ293; UNQ293/PRO332 | |
| Goat | |
| Antigen affinity chromatography | |
| RUO | |
| 242608 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Lyophilized |
| Immunohistochemistry (Frozen), Western Blot | |
| 0.2 mg/mL | |
| PBS with 5% trehalose and No Preservative | |
| Q7TQ62 | |
| Podn | |
| Mouse myeloma cell line NS0-derived recombinant mouse Podocan Val24-Arg611. | |
| 100 μg | |
| Primary | |
| Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction