Learn More
Brannan™ Pocket Dial Test Thermometer with 3 Point Traceable Calibration Certificate
Product Code. 17823098
Click to view available options
Calibration Points:
0°C, 50°C, 100°C
0°C, 50°C, 90°C
Unit Size:
Each
Product Code. 17823098
Brand: Brannan™ 31/143/0 + 49/503/0
Only null left
Add to basket
Add to basket
Pocket dial test thermometer with 24 mm diameter dial face with 125 mm spike stem. Complete with calibration.
  • bimetal dial with 125 mm stainless steel probe
  • Supplied with calibration certificate
  • Protective plastic pocket and case with clip
Specifications
Calibration Points 0°C, 50°C, 100°C
Certifications/Compliance ISO 17025 UKAS
Display Dimensions 24 mm dia.
For Use With (Application) To check the temperature
Length (Metric) 22.1 cm
Material Stainless Steel
Max. Temperature (English) -10°C
Max. Temperature (Metric) -10°C
Min. Temperature (English) 110°C
Min. Temperature (Metric) 110°C
Temperature Range (English) -10°C to +110°C
Temperature Range (Metric) -10°C to +110°C
Accuracy ±2% of scale
Subdivision 1.0°C
Immersion Length 76 mm