missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ PNCK Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA598524
This item is not returnable.
View return policy
Description
PNCK Polyclonal Antibody for Western Blot, IHC (P), ELISA
PNCK is a member of the calcium/calmodulin-dependent protein kinase family of protein serine/threonine kinases (Gardner et al., 2000 ).
Specifications
| PNCK | |
| Polyclonal | |
| Unconjugated | |
| PNCK | |
| BSTK3; calcium/calmodulin-dependent protein kinase 1 beta; calcium/calmodulin-dependent protein kinase 1, beta; calcium/calmodulin-dependent protein kinase type 1B; CaM kinase I beta; caM kinase IB; CaMK1b; caM-KI beta; caMKI-beta; CaMKIbeta2; caMKlb1; CaMKlb2; Pnck; pregnancy up-regulated nonubiquitous CaM kinase; pregnancy up-regulated non-ubiquitously expressed CaM kinase; pregnancy upregulated non-ubiquitously expressed CaM kinase; pregnancy up-regulated non-ubiquitously-expressed CaM kinase; Pregnancy up-regulated non-ubiquitously-expressed CaM kinase homolog; Punc | |
| Rabbit | |
| Protein G | |
| RUO | |
| 139728, 29660, 93843 | |
| -20°C or -80°C if preferred | |
| Liquid |
| ELISA, Immunohistochemistry (Paraffin), Western Blot | |
| 3.13 mg/mL | |
| PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4 | |
| O70150, Q6P2M8, Q9QYK9 | |
| PNCK | |
| Recombinant Human Calcium/calmodulin-dependent protein kinase type 1B protein (1-115AA). | |
| 100 μg | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction