missing translation for 'onlineSavingsMsg'
Learn More

PMS1 Mouse Monoclonal Antibody, Proteintech

Product Code. 30235825 Shop All Proteintech Products
Change view
Click to view available options
Quantity:
150 μL
20 μL
Förpackningsstorlek:
150µL
20µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Quantity unitSize
30235825 20 μL 20µL
30236093 150 μL 150µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Denna artikel kan inte returneras. Se returpolicy
Produktkod. 30235825 Leverantör Proteintech Leverantörsnummer 684131IG20UL

Please to purchase this item. Need a web account? Register with us today!

Denna artikel kan inte returneras. Se returpolicy

Mouse Monoclonal Antibody

The PMS1 antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human PMS1. This antibody recognizes human antigen. The PMS1 antibody has been validated for the following applications: WB, IF/ICC, ELISA analysis.

Specifikationer

Antigen PMS1
Applications Western Blot, Immunocytochemistry/Immunofluorescence, ELISA
Classification Monoclonal
Clone 1A6G3
Concentration 1000 μg/mL
Conjugate Unconjugated
Gene Symbols DKFZp781M0253; FLJ98259; HNPCC3; PMSL1; hPMS1
Host Species Mouse
Immunogen Recombinant Protein
Purification Method Protein A purification
Quantity 20 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 5378
Target Species Human
Content And Storage Store at -20°C. Stable for one year after shipment.
Antibody Molecular Weight 106 kDa
Product Type Monoclonal Antibody
Form Liquid
Isotype IgG2a
Visa mer Visa mindre
Produkttitel
Välj ett problem

Genom att klicka på Skicka bekräftar du att du kan bli kontaktad av Fisher Scientific angående feedbacken du har lämnat i detta formulär. Vi kommer inte att dela din information för andra ändamål. All kontaktinformation som tillhandahålls ska också underhållas i enlighet med vår Sekretesspolicy.