Learn More
Invitrogen™ PMF1 Recombinant Rabbit Monoclonal Antibody (24GB985)
GREENER_CHOICE
Product Code. 30275565
Change view
Click to view available options
Quantity:
20 μL
100 μL
Unit Size:
100µL
20µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30275565 100 μL 100µL
30276234 20 μL 20µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 30275565 Supplier Invitrogen™ Supplier No. MA557612
Rabbit Recombinant Monoclonal Antibody
PMF1 Recombinant Monoclonal Antibody for Western Blot, ICC/IF, Flow
PMF1 is part of the MIS12 complex which is required for normal chromosome alignment and segregation and kinetochore formation during mitosis. It may act as a cotranscription partner of NFE2L2 involved in regulation of polyamine-induced transcription of SSAT.
TRUSTED_SUSTAINABILITY
Specifications
Antigen PMF1
Applications Flow Cytometry, Western Blot, Immunocytochemistry
Classification Recombinant Monoclonal
Clone 24GB985
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene PMF1
Gene Accession No. Q6P1K2, Q9CPV5
Gene Alias 2600009M07Rik; AL033286; AW060657; Est1p-like protein B (EST1B); Pmf1; PMF-1; polyamine modulated factor 1; polyamine-modulated factor 1; RP11-54H19.4; Unknown (protein for MGC:134238)
Gene Symbols PMF1
Host Species Rabbit
Immunogen A synthesized peptide derived from human PMF1 (aa 1-205).
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 11243, 67037, 681050
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG