missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ PLXNB1 Monoclonal Antibody (439512)
GREENER_CHOICE

Product Code. 15583852
Change view
Click to view available options
Quantity:
100 μg
Unit Size:
100µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15583852 100 μg 100µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15583852 Supplier Invitrogen™ Supplier No. MA524165

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

In direct ELISAs and Western blots, no cross-reactivity with recombinant human (rh) Plexin C1, rhPlexin D1, recombinant mouse (rm) Plexin A1, rmPlexin A2, or rmPlexin A3 is observed. Reconstitute at 0.5 mg/mL in sterile PBS.

Plexin B1 is a protein of the plexin family that in humans is encoded by the PLXNB1gene. Receptor for SEMA4D. Plays a role in RHOA activation and subsequent changes of the actin cytoskeleton. Plays a role in axon guidance, invasive growth and cell migration.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PLXNB1
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Clone 439512
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 5% trehalose and No Preservative
Gene Plxnb1
Gene Accession No. O43157
Gene Alias 2900002G15Rik; AU042020; Kiaa0407; mKIAA0407; plexin 1; plexin 5; plexin B1; plexin-A1; PLEXIN-B1; PLXN5; Plxnb1; Semaphorin receptor SEP; SEP
Gene Symbols Plxnb1
Host Species Mouse
Immunogen E. coli-derived recombinant human Plexin B1 Leu35-Val150.
Purification Method Protein A/G
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 5364
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Lyophilized
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.