missing translation for 'onlineSavingsMsg'
Learn More

PLX7904, MedChemExpress

Product Code. 16407688
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16407688 10 mg 10mg
16487576 100 mg 100mg
16473687 50 mg 50mg
16420148 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 16407688 Supplier Medchem Express Supplier No. HY18997

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress PLX7904 is a potent and selective BRAF inhibitor, with IC50 of appr 5 nM against BRAFV600E in mutant RAS expressing cells.

PLX7904 is a potent and selective BRAF inhibitor, with IC50 of appr 5 nM against BRAFV600E in mutant RAS expressing cells.

Specifications

Chemical Name or Material PLX7904
CAS 1393465-84-3
Color Off-White
Purity Grade Notes Research
Molecular Formula C24H22F2N6O3S
Quantity 10 mg
Synonym PB04
Solubility Information DMSO : ≥ 30 mg/mL (58.53 mM)
SMILES O=S(NC1=CC=C(F)C(C(C2=CNC3=NC=C(C4=CN=C(C5CC5)N=C4)C=C32)=O)=C1F)(N(CC)C)=O
Molecular Weight (g/mol) 512.53
Formula Weight 512.53
Percent Purity 98.34%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-Kinase/protease
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.