Learn More
Kartell™ Pluripet II Variable Volume Micropipette
Click to view available options
Volume (Metric):
0.5, 5.0, 10.0 μL
100, 500, 1000 μL
Unit Size:
Each
Description
Specifications
Specifications
| Accuracy | ± 0,6 ± 0,3 ± 0,5 |
| Compatible Tips | Kartell™ 961 |
| Autoclavable | Autoclavable |
| Model | Pluripet II 1000 |
| Product Line | Pluripet II |
| Type | Micropipette |
| Volume (Metric) | 100, 500, 1000 μL |
| Increments | 10μL |