Learn More
Thermo Scientific™ Plumbing Adapter Kit for VersaCool™ Refrigerated Circulating Bath
Shop All Thermo Scientific Products
Click to view available options
Type:
Plumbing Adapter Kit
Unit Size:
Each
Description
Utilize the plumbing adapter kit to easily circulate the fluid from the bath to your application. Optimal performance with the Thermo Scientific™ VersaCool™ Refrigerated Circulating Bath.
A complete selection of hose barbs and threaded adapters are included as standard.
- 0.25 in. (8mm), 0.5 in (12mm) 8mm and 12mm hose barbs
- 0.25 in. MNPT and 0.5 in. MNPT – ideal for use with quick disconnects
Specifications
Specifications
| For Use With (Equipment) | Thermo Scientific VersaCool Refrigerated Circulating Bath |
| Type | Plumbing Adapter Kit |