Learn More
Invitrogen™ PLEKHA7 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5114002
Beschreibung
Immunogen sequence: PESRYQTLPG RGLSGSTSRL QQSSTIAPYV TLRRGLNAES SKATFPRPKS For short term use, store at 2-8°C up to 1 week. For long term storage, store at -20°C in small aliquots to prevent freeze-thaw cycles. Predicted homology: Cow: 93%; Dog: 100%; Guinea Pig: 100%; Horse: 100%; Human: 100%; Mouse: 100%; Rabbit: 100%; Rat: 93%.
PLEKHA7 is required for zonula adherens biogenesis and maintenance. Acts via its interaction with KIAA1543/Nezha, which anchors microtubules at their minus-ends to zonula adherens, leading to the recruitment of KIFC3 kinesin to the junctional site.
Spezifikation
| PLEKHA7 | |
| Polyclonal | |
| Unconjugated | |
| PLEKHA7 | |
| A430081P20Rik; AI414265; DKFZp686M22243; Hadp1; Heart adapter protein 1; PH domain-containing family A member 7; PH domain-containing family A member 7 antibody; pleckstrin homology domain containing A7; pleckstrin homology domain containing A7 antibody; pleckstrin homology domain containing, family A member 2; pleckstrin homology domain containing, family A member 7; pleckstrin homology domain-containing family A member 7; Plekha7 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 144100 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot | |
| 0.5 mg/mL | |
| PBS with 2% sucrose and 0.09% sodium azide | |
| Q6IQ23 | |
| PLEKHA7 | |
| Synthetic peptide directed towards the middle region of human PLEKHA7. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.