Learn More
PLAU Monoclonal Antibody (OTI1D4), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand:  Origene Technologies TA805314S
Encompass_Preferred
 View more versions of this product
Product Code. 17585203
  • 242.00 EUR / 30µL
Description
PLAU Monoclonal antibody specifically detects PLAU in Human samples. It is validated for Western Blot
Specifications
Specifications
PLAU
Monoclonal
1 mg/mL
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
Q14994
Mouse
Affinity chromatography
RUO
9970
-20° C, Avoid Freeze/Thaw Cycles
IgG1
Western Blot
OTI1D4
Unconjugated
NR1I3
antagonist of uPA; ATF; BDPLT5; CD87; HGNC:9052; plasminogen activator; plasminogen activator, urinary; plasminogen activator, urokinase; Plau; QPD; uPA; u-PA; UPAM; UPAR; U-plasminogen activator; Urinary plasminogen activator, urokinase; URK; URKR; Urokinase-type plasminogen activator; Urokinase-type plasminogen activator chain B; Urokinase-type plasminogen activator long chain A; Urokinase-type plasminogen activator short chain A
Human recombinant protein fragment corresponding to amino acids 1-236 of human NR1I3 produced in E.coli.
30 μL
Primary
Human
Liquid
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Promotions
Promotions