Learn More
VEE GEE Plato Hydrometer with Thermometer
Product Code. 17896573
Change view
Click to view available options
Brix Range:
0 to 32° Plato
0 to 8.5° Plato
15.5 to 24° Plato
7.5 to 16° Plato
Unit Size:
Each
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Brix Range unitSize
17896573 0 to 32° Plato Each
17876573 0 to 8.5° Plato Each
17846483 7.5 to 16° Plato Each
17866473 15.5 to 24° Plato Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Product Code. 17896573 Supplier VEE GEE Supplier No. 6614TS8
The Plato Hydrometer with Thermometer is calibrated to show the concentration of extract as a percentage by weight in brewery wort.
  • Calibration for use at 20°C/68°F
  • Thermometer filled with SafetyBLUE liquid
Specifications
Type Plato Hydrometer
Increments 0.5°
Length (Metric) 38 cm
Brix Range 0 to 32° Plato
Material Glass
Temperature Range (English) 30°F to 120°F
Temperature Range (Metric) -6.667°C to +54.4°C
Temperature of Standardization 20°C (68°F)