Learn More
Platinum Iridium foil, 0.05mm (0.002in) thick, 99.9% (metals basis excluding precious metals)
Product Code. 11374757
Click to view available options
Quantity:
100 x 100 mm
Unit Size:
Each
Product Code. 11374757
Brand: Thermo Scientific Alfa Aesar 041803.GH
Request Bulk or Custom Format
This item requires a price update prior to being ordered.
Request a Price for this item here.
Pt:Ir; 90:10 wt%
Platinum Iridium foil, 0.05mm (0.002in) thick, 99.9% (metals basis excluding precious metals) is used in non-standard thermocouples, electrical contacts and as electrodes in medical research.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Specifications
Quantity 100 x 100 mm
Molecular Formula Pt:Ir; 90:10 wt%
Percent Purity 99.9%