Learn More
Eppendorf™ Platforms for the Innova™ S44i Incubator Shaker
Click to view available options
Holds:
1 L Erlenmeyer Flasks
125 mL Erlenmeyer Flasks
2 L Erlenmeyer Flasks
2 L Lauber Tomson Flasks
2.8 L Fernbach Flasks
250 mL Erlenmeyer Flasks
4 L Erlenmeyer Flasks
500 mL Erlenmeyer Flasks
No. of Positions:
8
15
39
53
86
Unit Size:
Each
Description
- Holds Erlenmeyer, Fernbach or Lauber Thomson flasks
- Dedicated platform
Specifications
Specifications
| Holds | 2 L Lauber Tomson Flasks |
| For Use With (Equipment) | Innova™ S44i |
| No. of Positions | 15 |