Learn More
Labnet™ Platform for Orbit™ 300 Digital Multipurpose Shaker
Click to view available options
Holds:
Flasks, Bottles, etc.
Four Microplates
Plates, Dishes, Boxes, etc.
Model:
Flat Platform
Microplate Platform
Spring Loaded Platform
Unit Size:
Each
Description
- Three interchangeable platforms (available separately) accommodate a variety of sample containers to make the Orbit™ 300 a truly multipurpose shaker. The flat platform, covered with a nonslip rubber mat, holds plates, dishes, boxes, etc. The microplate platform will accommodate up to four microplates. The remaining platform has spring loaded retaining bars that adjust to hold flasks, bottles, racks and other containers in place.
- Three versatile platform options
- Shaking, mixing, agitating or vortexing
- Heavy-duty construction
- Safe for cold room or incubator use
Specifications
Specifications
| Holds | Plates, Dishes, Boxes, etc. |
| Product Type | Shaker Platform |
| Dimensions (L x W) | 30 x 30 cm |
| Model | Flat Platform |
| For Use With (Application) | Shaking Laboratory Vessels |
| For Use With (Equipment) | Orbit™ 300 Digital Multipurpose Shaker |
| Includes | Nonslip Rubber Mat |