Learn More
Thermo Scientific™ Plates and Modules with Affinity Binding Surfaces
Product Code. 10541493
Click to view available options
Color:
Black
Clear
White
Product Type:
Breakable Module F8 (x12) in Frame
Lock-Well Module C8 (x12) in Frame
Microplate
Solid Module F8 (x12) in Frame
Surface Treatment:
Glutathione
Streptavidin
Total Volume (Metric) Well:
120 μL
350 μL
400 μL
Unit Size:
Pack of 15
Pack of 5
Product Code. 10541493
Brand: Thermo Scientific™ 436016
Only null left
Add to basket
Add to basket
Help reduce variability in molecular orientation with these plates and modules with affinity binding surfaces.
Help reduce variability in molecular orientation with Thermo Scientific™ Plates and Modules with Affinity Binding Surfaces.
Affinity Capture Surfaces (Passive Streptavidin and Immobilizer Streptavidin, Nickel Chelate and Glutathione, BioBind):
  • Highly specific binding surfaces reduce variability in molecular orientation.
  • Immobilizer surfaces improve signal to noise ratios.
  • Streptavidin biotin interaction can be exploited to immobilize a wide range of biomolecules.
TRUSTED_SUSTAINABILITY
Specifications
Format Standard
Lid Without Lid
Detection Method Fluorescence
Array 8 x 12
Packaging 15 Cases
Color Black
Product Type Microplate
Material Polystyrene
Volume (Metric) Well 400 μL
No. of Wells 96
Volume (Metric) Working 100 μL
Sterility Non-sterile
Surface Treatment Streptavidin
Total Volume (Metric) Well 400 μL
Well Shape Round, Flat Bottom (F-well)
No. per Case 15
No. per Pack 1
Plate Blocking Unspecified
Quantity 15/Cs.
Target Molecule Biotinylated