Learn More
Kartell™ Plastilab™ Tapered Straight Connectors
BIOPHARMA
Product Code. 11934349
Change view
Click to view available options
Diameter (Metric):
4-6 mm
6-8 mm
8-10 mm
10-12 mm
12-14 mm
Packaging Quantity:
10/Pk.
Unit Size:
Pack of 10
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Diameter (Metric) Packaging Quantity unitSize
11934349 6-8 mm 10/Pk. Pack of 10
11924349 4-6 mm 10/Pk. Pack of 10
11944349 8-10 mm 10/Pk. Pack of 10
11954349 10-12 mm 10/Pk. Pack of 10
11974349 12-14 mm 10/Pk. Pack of 10
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Product Code. 11934349 Supplier Kartell™ Supplier No. 7051100
Edge
For straight connection of tubing with different diameters.
For straight connection of tubings with different diameters.
  • Constructed from polypropylene
  • Autoclavable
Specifications
Diameter (Metric) 6-8 mm
Autoclavable Autoclavable
Material Polypropylene
For Use With (Application) For straight connection of different diameter tubing
Packaging Quantity 10/Pk.
Port Size 3.5mm bore
Shape Straight
Product Type Straight Tapered Tubing Connector
Titolo del prodotto
Selezionare un problema
Facendo clic su Invia, l'utente riconosce che potrebbe essere contattato da Fisher Scientific in merito al feedback fornito in questo modulo. Non condivideremo le vostre informazioni per altri scopi. Tutte le informazioni di contatto fornite saranno conservate in conformità con la nostra Politica sulla privacy. Informativa sulla privacy.