Learn More
Kartell™ Plastilab™ Tapered Straight Connectors
Product Code. 11924349
Change view
Click to view available options
Diameter (Metric):
4-6 mm
6-8 mm
8-10 mm
10-12 mm
12-14 mm
Packaging Quantity:
10/Pk.
Unit Size:
Pack of 10
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Diameter (Metric) Packaging Quantity unitSize
11924349 4-6 mm 10/Pk. Pack of 10
11934349 6-8 mm 10/Pk. Pack of 10
11944349 8-10 mm 10/Pk. Pack of 10
11954349 10-12 mm 10/Pk. Pack of 10
11974349 12-14 mm 10/Pk. Pack of 10
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Product Code. 11924349 Supplier Kartell™ Supplier No. 7051000
For straight connection of tubing with different diameters.
For straight connection of tubings with different diameters.
  • Constructed from polypropylene
  • Autoclavable
Specifications
Diameter (Metric) 4-6 mm
Autoclavable Autoclavable
Material Polypropylene
For Use With (Application) For straight connection of different diameter tubing
Packaging Quantity 10/Pk.
Port Size 2mm bore
Shape Straight
Product Type Straight Tapered Tubing Connector