Learn More
KD Scientific Plastic Syringe
Click to view available options
Volume (Metric):
1 mL
3 mL
5 mL
10 mL
20 mL
30 mL
Connection Type:
Luer Lock
Luer Slip
Tuberculin Luer Slip
Unit Size:
Pack of 25
Description
- A new line of disposable plastic sterile syringes for all scientific applications
- Norm-ject syringes are the ideal solution for any situation
- Their unique two-part system is latex free and contains no silicone lubricant or rubber
Specifications
Specifications
| Material | Polyethylene (Plunger) |
| Needle Gauge | No Needle Included |
| Latex Free | Yes |
| Product Type | Syringe |
| Disposable | Single-use |
| Volume (Metric) | 30 mL |
| For Use With (Equipment) | KD Scientific Syringe Pumps |
| Connection Type | Luer Slip |
| Graduated | Yes |
| Sterility | Sterile |
Korrektion af produktindhold
Dit input er vigtigt for os. Udfyld denne formular for at give feedback relateret til indholdet på dette produkt.
Produkttitel
Ser du en mulighed for forbedring?Del en indholdskorrektion